Assay Details
| Gene Symbol: Pnpla8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750150.3 | XP_003750198.1 | 9 - 10 | 2277 | 79 | |
| GenBank mRNA | FQ211866.1 | - | 4 - 5 | 486 | 79 | |
| FQ224093.1 | - | 3 - 4 | 457 | 79 | ||
More Information
Set Membership: |
|

Pnpla8
Genomic Map