Assay Details
| Gene Symbol: Slc38a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013099.1 | NP_001013117.1 | 10 - 11 | 807 | 117 | |
| XM_006240209.1 | XP_006240271.1 | 10 - 11 | 797 | 117 | ||
| XM_006240210.2 | XP_006240272.1 | 10 - 11 | 830 | 117 | ||
| GenBank mRNA | AY266018.1 | - | 10 - 11 | 807 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P030704 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P149267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9924 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P030704 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P149267 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P149267 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P030704 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P030704 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P149267 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P149267 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P149273 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P295478 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43928 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 87955 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30808 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46201 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74600 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10885234 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885226 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885233 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885224 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885231 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885222 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885225 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885223 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885221 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885220 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885228 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885230 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885218 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885232 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885227 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885219 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10885217 | - | - |
| Rat Genome 230 2.0 Array | 1399089_at | - | - |
| Rat Genome 230 2.0 Array | 1399089_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1399089_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1399089_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600006624.rn.1 | TC0600000974.rn.1 | RTA TC |
| Rat Expression Set 230 | 1399089_at | - | - |
| Rat Expression Set 230 | 1399089_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885217 | - | - |
| Rat Genome U34 Set | rc_AI072555_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21473783 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373638 | - | - |

Slc38a6
Genomic Map