Assay Details
| Gene Symbol: Mbip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108712.2 | NP_001102182.2 | 1 - 2 | 201 | 100 | |
| XM_006240120.2 | XP_006240182.1 | 1 - 2 | 219 | 100 | ||
| GenBank mRNA | BC166982.1 | - | 1 - 2 | 128 | 100 | |
| CB609032.1 | - | 1 - 2 | 207 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mbip
Genomic Map