Assay Details
| Gene Symbol: Prkar2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030020.1 | NP_001025191.1 | 7 - 8 | 840 | 74 | |
| GenBank mRNA | BC099223.1 | - | 6 - 7 | 583 | 7388087 | 74 |
| M12492.1 | - | 7 - 8 | 834 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid metabolic process
learning
regulation of protein kinase activity
response to antipsychotic drug
response to clozapine
negative regulation of cAMP-dependent protein kinase activity
cAMP-dependent protein kinase inhibitor activity
protein binding
cAMP-dependent protein kinase regulator activity
protein domain specific binding
cAMP binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding
fatty acid metabolic process
learning
regulation of protein kinase activity
response to antipsychotic drug
response to clozapine
negative regulation of cAMP-dependent protein kinase activity
cAMP-dependent protein kinase inhibitor activity
protein binding
cAMP-dependent protein kinase regulator activity
protein domain specific binding
cAMP binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P622681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143462 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P622681 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P622681 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P622681 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P116634 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P316122 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97336 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52972 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93709 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39872 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 28021 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10889596 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889591 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889599 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889598 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889600 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889594 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889601 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889602 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889592 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889603 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889604 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889595 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889593 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889597 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10889590 | - | - |
| Rat Genome 230 2.0 Array | 1371133_a_at | - | - |
| Rat Genome 230 2.0 Array | 1371133_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1371133_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1371133_PM_a_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600016547.rn.1 | TC0600002283.rn.1 | RTA TC |
| Rat Expression Set 230 | 1371133_PM_a_at | - | - |
| Rat Expression Set 230 | 1371133_a_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889590 | - | - |
| Rat Genome U34 Set | rc_AI235758_s_at | - | - |
| Rat Genome U34 Set | M12492mRNA#1_at | - | - |
| Rat Genome U34 Set | M12492mRNA#1_g_at | - | - |
| Rat Genome U34 Set | rc_AI030779_f_at | - | - |
| Rat Genome U34 Set | AA801167_i_at | - | - |
| Rat Genome U34 Set | M12492mRNA#1_g_at | - | - |
| Rat Genome U34 Set | M12492mRNA#1_at | - | - |
| Rat Genome U34 Set | rc_AI235758_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21156288 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353803 | - | - |

Prkar2b
Genomic Map