Assay Details
| Gene Symbol: Rbks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108703.1 | NP_001102173.1 | 2 - 3 | 256 | 145 | |
| XM_006239805.2 | XP_006239867.1 | 2 - 3 | 302 | 145 | ||
| XM_006239806.2 | XP_006239868.1 | 2 - 3 | 302 | 145 | ||
| XM_008764520.1 | XP_008762742.1 | 2 - 3 | 750 | 145 | ||
| XM_008764521.1 | XP_008762743.1 | 2 - 3 | 302 | 145 | ||
| GenBank mRNA | FQ209797.1 | - | 2 - 3 | 238 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbks
Genomic Map