Assay Details
| Gene Symbol: RGD1565002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271394.1 | NP_001258323.1 | 448 | 62 | ||
| GenBank mRNA | CV080419.1 | - | 3 - 4 | 408 | 62 | |
| FM087146.1 | - | 2 - 3 | 222 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RGD1565002
Genomic Map