Assay Details
| Gene Symbol: RGD1565002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271394.1 | NP_001258323.1 | 685 | 72 | ||
| GenBank mRNA | CV080419.1 | - | 4 - 5 | 645 | 72 | |
| FM087146.1 | - | 3 - 4 | 459 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RGD1565002
Genomic Map