Assay Details
| Gene Symbol: Preb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170708.1 | NP_001164179.1 | 1 - 2 | 306 | 67 | |
| GenBank mRNA | BC078936.1 | - | 1 - 2 | 306 | 7108249 | 67 |
More Information
Updated Mapping Information
Set Membership: |
|

Preb
Genomic Map