Assay Details
| Gene Symbol: Agr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106725.1 | NP_001100195.1 | 5 - 6 | 556 | 70 | |
| XM_006240041.2 | XP_006240103.1 | 5 - 6 | 558 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agr2
Genomic Map