Assay Details
| Gene Symbol: Trappc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239964.2 | XP_006240026.1 | 2 - 3 | 1351 | 63 | |
| XM_234029.8 | XP_234029.3 | 2 - 3 | 1383 | 63 | ||
| GenBank mRNA | FQ225443.1 | - | 3 - 4 | 1278 | 63 | |
| FQ226500.1 | - | 3 - 4 | 1278 | 63 | ||
More Information
Set Membership: |
|

Trappc12
Genomic Map