Assay Details
| Gene Symbol: Car8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009662.1 | NP_001009662.1 | 6 - 7 | 702 | 149 | |
| GenBank mRNA | BC087586.1 | - | 6 - 7 | 702 | 7318288 | 149 |
More Information
Updated Mapping Information
Set Membership: |
|

Car8
Genomic Map