Assay Details
| Gene Symbol: Car8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009662.1 | NP_001009662.1 | 7 - 8 | 812 | 91 | |
| GenBank mRNA | BC087586.1 | - | 7 - 8 | 812 | 7318288 | 91 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P124354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79179 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19656 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P124354 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P124354 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P124354 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P319080 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101512 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97334 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 83325 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37477 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30662 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77178 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29227 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78185 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10875381 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875380 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875377 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875382 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875384 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875383 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875376 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875378 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875379 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10875375 | - | - |
| Rat Genome 230 2.0 Array | 1398431_at | - | - |
| Rat Genome 230 2.0 Array | 1398431_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1398431_at | - | - |
| Affymetrix HT Rat Focus Array | 1398431_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500017267.rn.1 | TC0500002279.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500017268.rn.1 | TC0500002279.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500017266.rn.1 | TC0500002279.rn.1 | RTA TC |
| Rat Expression Set 230 | 1398431_PM_at | - | - |
| Rat Expression Set 230 | 1398431_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875375 | - | - |
| Rat Genome U34 Set | rc_AI100768_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369426 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1530384 | - | - |
| RatRef-12 Expression BeadChip | ILMN_52247 | - | - |
| RatRef-12 Expression BeadChip | ILMN_161401 | - | - |

Car8
Genomic Map