Assay Details
| Gene Symbol: Rlim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024892.1 | NP_001020063.1 | 1 - 2 | 119 | 167 | |
| XM_006257149.2 | XP_006257211.1 | 1 - 2 | 202 | 279 | ||
| GenBank mRNA | BC097491.1 | - | 1 - 2 | 119 | 7461011 | 167 |
| Gene Symbol: LOC100910646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003752081.3 | XP_003752129.1 | 202 | 167 | ||
| XM_006257141.2 | XP_006257203.1 | 202 | 279 | |||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC097491.1,NM_001024892.1,XM_003752081.3,XM_006257141.2,XM_006257149.2 transcripts.
Set Membership: |
|

Rlim
Genomic Map