Assay Details
| Gene Symbol: LOC103690008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257157.2 | XP_006257219.2 | 747 | 152 | ||
| XM_008773351.1 | XP_008771573.1 | 747 | 152 | |||
| XM_008773352.1 | XP_008771574.1 | 2 - 3 | 747 | 152 | ||
| XM_008773353.1 | XP_008771575.1 | 2 - 3 | 747 | 152 | ||
| Gene Symbol: Atrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773366.1 | XP_008771588.1 | 4 - 5 | 407 | 152 | |
More Information
Additional Information:
				    	 
					    	
					        	
					            	This assay is in the 5'UTR region of the XM_006257157.2,XM_008773351.1,XM_008773352.1,XM_008773353.1 transcripts.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
					        	
					    	            
				    	
			    	
			    
			Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: | 
		   
					
						
							 | 
				

LOC103690008
Genomic Map