Assay Details
| Gene Symbol: Lpar4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106940.1 | NP_001100410.1 | 2 - 3 | 472 | 113 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106940.1 transcripts.
Set Membership: |
|

Lpar4
Genomic Map