Assay Details
| Gene Symbol: Fam46d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106936.1 | NP_001100406.1 | 1 | 1232 | 157 | |
| XM_006257181.2 | XP_006257243.1 | 4 | 1647 | 157 | ||
| XM_008773335.1 | XP_008771557.1 | 5 | 1759 | 157 | ||
| XM_008773336.1 | XP_008771558.1 | 5 | 1525 | 157 | ||
| XM_008773337.1 | XP_008771559.1 | 5 | 1973 | 157 | ||
| XM_008773338.1 | XP_008771560.1 | 4 | 1645 | 157 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fam46d
Genomic Map