Assay Details
| Gene Symbol: Doc2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011937.1 | NP_001011937.1 | 4 - 5 | 533 | 120 | |
| GenBank mRNA | BC083779.1 | - | 4 - 5 | 533 | 7193879 | 120 |
More Information
Updated Mapping Information
Set Membership: |
|

Doc2g
Genomic Map