Assay Details
| Gene Symbol: Doc2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011937.1 | NP_001011937.1 | 8 - 9 | 989 | 95 | |
| XM_006230735.2 | XP_006230797.1 | 8 - 9 | 2013 | 95 | ||
| GenBank mRNA | BC083779.1 | - | 8 - 9 | 989 | 7193879 | 95 |
More Information
Updated Mapping Information
Set Membership: |
|

Doc2g
Genomic Map