Assay Details
| Gene Symbol: LOC100910732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751066.2 | XP_003751114.1 | 4 - 5 | 716 | 115 | |
| XM_006248473.1 | XP_006248535.1 | 5 - 6 | 845 | 115 | ||
| XM_008768778.1 | XP_008767000.1 | 4 - 5 | 2277 | 115 | ||
| Gene Symbol: Pak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053306.3 | NP_445758.2 | 568 | 115 | ||
| GenBank mRNA | S80221.1 | - | 487 | 115 | ||
| U19967.1 | - | 568 | 115 | |||
| U35345.1 | - | 487 | 115 | |||
More Information
Updated Mapping Information
Set Membership: |
|

LOC100910732
Genomic Map