Assay Details
| Gene Symbol: LOC100910732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751066.2 | XP_003751114.1 | 7 - 8 | 985 | 143 | |
| XM_006248473.1 | XP_006248535.1 | 8 - 9 | 1114 | 143 | ||
| XM_008768778.1 | XP_008767000.1 | 7 - 8 | 2546 | 143 | ||
| Gene Symbol: Pak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053306.3 | NP_445758.2 | 837 | 143 | ||
| GenBank mRNA | S80221.1 | - | 756 | 143 | ||
| U19967.1 | - | 837 | 143 | |||
| U35345.1 | - | 756 | 143 | |||
More Information
Updated Mapping Information
Set Membership: |
|

LOC100910732
Genomic Map