Assay Details
| Gene Symbol: Cd80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012926.1 | NP_037058.1 | 1 - 2 | 118 | 68 | |
| XM_006248365.2 | XP_006248427.1 | 1 - 2 | 958 | 68 | ||
| GenBank mRNA | AF010465.1 | - | 1 - 2 | 118 | 68 | |
| U10925.1 | - | 1 - 2 | 392 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd80
Genomic Map