Assay Details
| Gene Symbol: C2cd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199391.1 | NP_955423.1 | 5 - 6 | 939 | 63 | |
| XM_008768572.1 | XP_008766794.1 | 5 - 6 | 1046 | 63 | ||
| XM_008768573.1 | XP_008766795.1 | 5 - 6 | 1043 | 63 | ||
| XM_008768574.1 | XP_008766796.1 | 5 - 6 | 1049 | 63 | ||
| XM_008768575.1 | XP_008766797.1 | 5 - 6 | 1049 | 63 | ||
| XM_008768576.1 | XP_008766798.1 | 6 - 7 | 648 | 63 | ||
| XM_008768577.1 | XP_008766799.1 | 5 - 6 | 523 | 63 | ||
| XM_008768578.1 | XP_008766800.1 | 5 - 6 | 886 | 63 | ||
| GenBank mRNA | BC062389.1 | - | 5 - 6 | 939 | 5620907 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

C2cd2
Genomic Map