Assay Details
| Gene Symbol: Osbpl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107090.2 | NP_001100560.1 | 7 - 8 | 1478 | 70 | |
| XM_006248450.2 | XP_006248512.1 | 6 - 7 | 1533 | 70 | ||
| XM_006248451.2 | XP_006248513.1 | 7 - 8 | 1109 | 70 | ||
| XM_006248452.2 | XP_006248514.1 | 7 - 8 | 990 | 70 | ||
| XM_008768726.1 | XP_008766948.1 | 7 - 8 | 1259 | 70 | ||
| GenBank mRNA | BC168952.1 | - | 7 - 8 | 1478 | 7453666 | 70 |
More Information
Updated Mapping Information
Set Membership: |
|

Osbpl11
Genomic Map