Assay Details
| Gene Symbol: Tfg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012144.1 | NP_001012144.1 | 1 - 2 | 220 | 122 | |
| XM_006248236.2 | XP_006248298.1 | 1 - 2 | 346 | 122 | ||
| XM_006248237.2 | XP_006248299.1 | 1 - 2 | 346 | 122 | ||
| GenBank mRNA | BC078947.1 | - | 1 - 2 | 220 | 7109719 | 122 |
| Gene Symbol: LOC103690095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768670.1 | XP_008766892.1 | 356 | 122 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC078947.1, NM_001012144.1, XM_006248236.2, XM_006248237.2, XM_008768670.1 transcripts.
Set Membership: |
|

Tfg
Genomic Map