Assay Details
| Gene Symbol: Tfg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012144.1 | NP_001012144.1 | 2 - 3 | 447 | 109 | |
| XM_006248236.2 | XP_006248298.1 | 2 - 3 | 573 | 109 | ||
| XM_006248237.2 | XP_006248299.1 | 2 - 3 | 573 | 109 | ||
| GenBank mRNA | AB218900.1 | - | 1 - 2 | 188 | 109 | |
| BC078947.1 | - | 2 - 3 | 447 | 7109719 | 109 | |
| Gene Symbol: LOC103690095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768670.1 | XP_008766892.1 | 583 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tfg
Genomic Map