Assay Details
| Gene Symbol: Bcorl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191587.1 | NP_001178516.1 | 10 - 11 | 4903 | 59 | |
| XM_006257523.2 | XP_006257585.1 | 10 - 11 | 6377 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bcorl1
Genomic Map