Assay Details
| Gene Symbol: RGD1564541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107950.1 | NP_001101420.1 | 2 - 3 | 379 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564541| Gene Symbol: RGD1564541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107950.1 | NP_001101420.1 | 2 - 3 | 379 | 87 | |
Set Membership: |
|