Assay Details
| Gene Symbol: Vrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005561.1 | NP_001005561.1 | 10 - 11 | 990 | 82 | |
| XM_006229095.2 | XP_006229157.1 | 10 - 11 | 1049 | 82 | ||
| XM_006229096.2 | XP_006229158.1 | 10 - 11 | 1050 | 82 | ||
| GenBank mRNA | BC081928.1 | - | 10 - 11 | 990 | 7113277 | 82 |
More Information
Updated Mapping Information
Set Membership: |
|

Vrk3
Genomic Map