Assay Details
| Gene Symbol: Yif1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014810.1 | NP_001014810.1 | 4 - 5 | 507 | 76 | |
| NM_198734.2 | NP_942029.2 | 4 - 5 | 517 | 76 | ||
| XM_008759128.1 | XP_008757350.1 | 4 - 5 | 536 | 76 | ||
| XM_008759129.1 | XP_008757351.1 | 4 - 5 | 516 | 76 | ||
| GenBank mRNA | BC058153.1 | - | 4 - 5 | 510 | 6887903 | 76 |
| CB576820.1 | - | 4 - 5 | 460 | 76 | ||
| CB751111.1 | - | 1 - 2 | 113 | 76 | ||
| Gene Symbol: LOC103689986 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228658.2 | XP_006228720.1 | 4 - 5 | 536 | 76 | |
| XM_006228660.2 | XP_006228722.1 | 4 - 5 | 516 | 76 | ||
| XM_008759221.1 | XP_008757443.1 | 4 - 5 | 546 | 76 | ||
| XM_008759222.1 | XP_008757444.1 | 4 - 5 | 565 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Yif1b
Genomic Map