Assay Details
| Gene Symbol: Exoc3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228497.2 | XP_006228559.1 | 9 - 10 | 1889 | 82 | |
| XM_006228498.2 | XP_006228560.1 | 9 - 10 | 2025 | 82 | ||
More Information
Set Membership: |
|

Exoc3l2
Genomic Map