Assay Details
| Gene Symbol: LOC100910371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244579.2 | XP_006244641.1 | 1 - 2 | 161 | 89 | |
| Gene Symbol: Dmrtc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109140.1 | NP_001102610.1 | 1 - 2 | 107 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109140.1, XM_006244579.2 transcripts.
Set Membership: |
|

LOC100910371
Genomic Map