Assay Details
| Gene Symbol: Scn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271045.1 | NP_001257974.1 | 4 - 5 | 731 | 107 | |
| NM_001271046.2 | NP_001257975.1 | 4 - 5 | 598 | 107 | ||
| NM_017288.2 | NP_058984.1 | 4 - 5 | 812 | 107 | ||
| GenBank mRNA | BC094523.1 | - | 4 - 5 | 731 | 7316686 | 107 |
| FQ215379.1 | - | 2 - 3 | 389 | 107 | ||
| FQ216374.1 | - | 4 - 5 | 598 | 107 | ||
| M91808.1 | - | 4 - 5 | 811 | 107 | ||
More Information
Set Membership: |
|

Scn1b
Genomic Map