Assay Details
| Gene Symbol: Lrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053541.1 | NP_445993.1 | 1 - 2 | 443 | 62 | |
| XM_008759200.1 | XP_008757422.1 | 1 - 2 | 522 | 62 | ||
| XM_008759202.1 | XP_008757424.1 | 1 - 2 | 522 | 62 | ||
| GenBank mRNA | AB009463.1 | - | 1 - 2 | 443 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrp3
Genomic Map