Assay Details
| Gene Symbol: Ceacam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033860.1 | NP_001029032.1 | 3 - 4 | 764 | 130 | |
| NM_001033861.1 | NP_001029033.1 | 3 - 4 | 764 | 130 | ||
| NM_001033862.1 | NP_001029034.1 | 3 - 4 | 764 | 130 | ||
| NM_031755.2 | NP_113943.1 | 3 - 4 | 764 | 130 | ||
| GenBank mRNA | AJ277104.1 | - | 3 - 4 | 726 | 130 | |
| AJ277105.1 | - | 3 - 4 | 726 | 130 | ||
| J04963.1 | - | 3 - 4 | 771 | 130 | ||
| M92848.1 | - | 3 - 4 | 764 | 130 | ||
| X91137.1 | - | 3 - 4 | 726 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ceacam1
Genomic Map