Assay Details
| Gene Symbol: Pvrl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012064.1 | NP_001012064.1 | 7 - 8 | 1451 | 59 | |
| GenBank mRNA | BC090075.1 | - | 7 - 8 | 1451 | 7189358 | 59 |
More Information
Updated Mapping Information
Set Membership: |
|

Pvrl2
Genomic Map