Assay Details
| Gene Symbol: Ceacam18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003748856.3 | XP_003748904.2 | 1 - 2 | 392 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ceacam18| Gene Symbol: Ceacam18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003748856.3 | XP_003748904.2 | 1 - 2 | 392 | 106 | |
Set Membership: |
|