Assay Details
| Gene Symbol: Fuz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037646.1 | NP_001032735.1 | 4 - 5 | 522 | 78 | |
| XM_006229059.2 | XP_006229121.1 | 4 - 5 | 561 | 78 | ||
| XM_006229060.2 | XP_006229122.1 | 4 - 5 | 561 | 78 | ||
| XM_006229061.2 | XP_006229123.1 | 4 - 5 | 614 | 78 | ||
| XM_008759373.1 | XP_008757595.1 | 4 - 5 | 562 | 78 | ||
| XM_008759374.1 | XP_008757596.1 | 4 - 5 | 562 | 78 | ||
| GenBank mRNA | BC107809.1 | - | 4 - 5 | 522 | 7453736 | 78 |
| FQ228777.1 | - | 4 - 5 | 507 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fuz
Genomic Map