Assay Details
| Gene Symbol: Snrpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008303.1 | NP_001008304.1 | 5 - 6 | 680 | 63 | |
| XM_006228564.2 | XP_006228626.1 | 5 - 6 | 811 | 63 | ||
| GenBank mRNA | BC086331.1 | - | 5 - 6 | 680 | 7300752 | 63 |
| FQ227089.1 | - | 4 - 5 | 574 | 63 | ||
| FQ231860.1 | - | 3 - 4 | 384 | 63 | ||
| FQ233559.1 | - | 4 - 5 | 574 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Snrpa
Genomic Map