Assay Details
Gene Symbol: Emc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004221.1 | NP_001004221.1 | 2 - 3 | 266 | 62 | |
XM_006229000.2 | XP_006229062.1 | 2 - 3 | 282 | 62 | ||
XM_006229001.2 | XP_006229063.1 | 2 - 3 | 282 | 62 | ||
GenBank mRNA | BC079041.1 | - | 2 - 3 | 266 | 7114835 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
