Assay Details
| Gene Symbol: Psenen | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228770.2 | XP_006228832.1 | 1 - 2 | 78 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006228770.2 transcripts.
Set Membership: |
|

Psenen
Genomic Map