Assay Details
| Gene Symbol: B3gnt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107492.1 | NP_001100962.1 | 1 - 2 | 226 | 77 | |
| XM_006228436.2 | XP_006228498.1 | 1 - 2 | 924 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107492.1, XM_006228436.2 transcripts.
Set Membership: |
|

B3gnt8
Genomic Map