Assay Details
| Gene Symbol: Rabac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031774.1 | NP_113962.1 | 2 - 3 | 364 | 60 | |
| GenBank mRNA | AF025506.1 | - | 2 - 3 | 364 | 60 | |
| BC086387.1 | - | 2 - 3 | 331 | 7302383 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rabac1
Genomic Map