Assay Details
| Gene Symbol: Klc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138520.1 | NP_612529.1 | 6 - 7 | 890 | 133 | |
| XM_006228377.2 | XP_006228439.1 | 6 - 7 | 1043 | 133 | ||
| XM_006228378.2 | XP_006228440.1 | 6 - 7 | 1043 | 133 | ||
| XM_006228379.2 | XP_006228441.1 | 6 - 7 | 965 | 133 | ||
| XM_006228380.2 | XP_006228442.1 | 6 - 7 | 965 | 133 | ||
| GenBank mRNA | AF166267.1 | - | 6 - 7 | 890 | 133 | |
| BC078736.1 | - | 6 - 7 | 903 | 7134584 | 133 | |
| BC097284.1 | - | 6 - 7 | 895 | 7454436 | 133 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klc3
Genomic Map