Assay Details
| Gene Symbol: Klc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138520.1 | NP_612529.1 | 8 - 9 | 1173 | 110 | |
| XM_006228377.2 | XP_006228439.1 | 8 - 9 | 1326 | 110 | ||
| XM_006228378.2 | XP_006228440.1 | 8 - 9 | 1326 | 110 | ||
| XM_006228379.2 | XP_006228441.1 | 8 - 9 | 1248 | 110 | ||
| XM_006228380.2 | XP_006228442.1 | 8 - 9 | 1248 | 110 | ||
| GenBank mRNA | AF166267.1 | - | 8 - 9 | 1173 | 110 | |
| BC078736.1 | - | 8 - 9 | 1186 | 7134584 | 110 | |
| BC097284.1 | - | 8 - 9 | 1178 | 7454436 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klc3
Genomic Map