Assay Details
| Gene Symbol: Hdgfrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145785.1 | NP_665728.1 | 3 - 4 | 421 | 128 | |
| XM_008759496.1 | XP_008757718.1 | 3 - 4 | 696 | 128 | ||
| GenBank mRNA | AF389347.1 | - | 3 - 4 | 421 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hdgfrp3
Genomic Map