Assay Details
| Gene Symbol: Ap3s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115039.1 | NP_001108511.1 | 1 - 2 | 200 | 107 | |
| GenBank mRNA | BC158881.1 | - | 1 - 2 | 200 | 7936771 | 107 |
More Information
Updated Mapping Information
Set Membership: |
|

Ap3s2
Genomic Map