Assay Details
| Gene Symbol: Isg20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008510.1 | NP_001008510.1 | 1 - 2 | 266 | 60 | |
| GenBank mRNA | BC086557.1 | - | 1 - 2 | 266 | 7300717 | 60 |
| FQ233960.1 | - | 2 - 3 | 323 | 60 | ||
| FQ234031.1 | - | 333 | 60 | |||
| FQ234316.1 | - | 1 - 2 | 319 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Isg20
Genomic Map