Assay Details
| Gene Symbol: Cpeb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106276.1 | NP_001099746.1 | 3 - 4 | 407 | 59 | |
| XM_008759509.1 | XP_008757731.1 | 3 - 4 | 1016 | 59 | ||
| XM_008759510.1 | XP_008757732.1 | 3 - 4 | 1016 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpeb1
Genomic Map