Assay Details
| Gene Symbol: Mctp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191625.1 | NP_001178554.1 | 9 - 10 | 1280 | 67 | |
| XM_006229375.2 | XP_006229437.1 | 10 - 11 | 1629 | 67 | ||
| XM_008759522.1 | XP_008757744.1 | 10 - 11 | 1630 | 67 | ||
| XM_008759523.1 | XP_008757745.1 | 10 - 11 | 1958 | 67 | ||
| XM_008759524.1 | XP_008757746.1 | 11 - 12 | 1703 | 67 | ||
| XM_008759525.1 | XP_008757747.1 | 3 - 4 | 739 | 67 | ||
| XM_008759526.1 | XP_008757748.1 | 2 - 3 | 139 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mctp2
Genomic Map