Assay Details
| Gene Symbol: Ppapdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109411.1 | NP_001102881.2 | 5 - 6 | 490 | 105 | |
| XM_008771367.1 | XP_008769589.1 | 5 - 6 | 968 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppapdc1b
Genomic Map